Merck
  • A spectrophotometric method for the estimation of polymer-supported sulfhydryl groups.
A spectrophotometric method for the estimation of polymer-supported sulfhydryl groups.
Analytical biochemistry (1990-09-01)
P Sharma, S Sathyanarayana, P Kumar, K C Gupta
ABSTRACT
A sensitive and simple method is described for the quantitative determination of free sulfhydryl (-SH) groups on polymer supports. The method includes the reaction of 4,4'-dimethoxytrityloxy-S-(2-thio-5-nitropyridyl)-2-mercapto ethane (DTNPME) with polymer-supported sulfhydryl groups. After removal of excess reagent through washing, a weighed quantity of the polymer support is treated with perchloric acid to release the 4,4'-dimethoxytrityl cation from the polymer support into the solution. The dimethoxytrityl cation (lambda max = 498 nm, epsilon 498 = 70,000/M) is then quantified spectrophotometrically. A comparative study of the reagent DTNPME with 2,2'-dithiobis(5-nitropyridine) is also described.
Sigma-Aldrich
2,2′-Dithiobis(5-nitropyridine), 96%
Pricing and availability is not currently available.