Merck
Inverse rectification in donor-acceptor molecular heterojunctions.
ACS nano (2011-10-21)
Shannon K Yee, Jibin Sun, Pierre Darancet, T Don Tilley, Arun Majumdar, Jeffrey B Neaton, Rachel A Segalman
ABSTRACT
The transport properties of a junction consisting of small donor-acceptor molecules bound to Au electrodes are studied and understood in terms of its hybrid donor-acceptor-electrode interfaces. A newly synthesized donor-acceptor molecule consisting of a bithiophene donor and a naphthalenediimide acceptor separated by a conjugated phenylacetylene bridge and a nonconjugated end group shows rectification in the reverse polarization, behavior opposite to that observed in mesoscopic p-n junctions. Solution-based spectroscopic measurements demonstrate that the molecule retains many of its original constituent properties, suggesting a weak hybridization between the wave functions of the donor and acceptor moieties, even in the presence of a conjugated bridge. Differential conductance measurements for biases as high as 1.5 V are reported and indicate a large asymmetry in the orbital contributions to transport arising from disproportionate electronic coupling at anode-donor and acceptor-cathode interfaces. A semi-empirical single Lorentzian coherent transport model, developed from experimental data and density functional theory based calculations, is found to explain the inverse rectification.
Sigma-Aldrich
Phenylacetylene, 98%