Merck
  • Synthesis of functionalized 1H-indenes via copper-catalyzed arylative cyclization of arylalkynes with aromatic sulfonyl chlorides.
Synthesis of functionalized 1H-indenes via copper-catalyzed arylative cyclization of arylalkynes with aromatic sulfonyl chlorides.
Journal of the American Chemical Society (2011-10-13)
Xiaoming Zeng, Laurean Ilies, Eiichi Nakamura
ABSTRACT
A variety of polysubstituted 1H-indenes can be prepared through the copper-catalyzed arylative cyclization of simple arylalkynes with commercially available aromatic sulfonyl chlorides that function as an aryl group donor. The reaction tolerates a broad range of functional groups, including bromide and iodide, nitrile, ketone, and nitro groups. The reaction allowed the synthesis of polycyclic aromatic hydrocarbons, such as a bis(indene), indacene, and fused polyarene derivatives, some of them showing strong fluorescence in solution and the solid state.
Sigma-Aldrich
Sulfuryl chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Sulfuryl chloride, 97%