Merck
  • Prospective randomized controlled trial of combination ranibizumab (Lucentis) and bromfenac (Xibrom) for neovascular age-related macular degeneration: a pilot study.
Prospective randomized controlled trial of combination ranibizumab (Lucentis) and bromfenac (Xibrom) for neovascular age-related macular degeneration: a pilot study.
Retina (Philadelphia, Pa.) (2011-08-25)
Christina Flaxel, Mitchell B Schain, Sara C Hamon, Peter J Francis
ABSTRACT
To evaluate whether bromfenac eyedrops and ranibizumab intravitreal injections would provide added efficacy over ranibizumab alone. This was a single-site, multiinvestigator, prospective, open-label, interventional, Phase II study of patients with new or recurrent exudative/neovascular age-related macular degeneration. Thirty eyes were enrolled consecutively and were randomized in a ratio of 2:1 to combination therapy with intravitreal ranibizumab and topical bromfenac, and ranibizumab alone. All patients received ranibizumab monthly therapy for 4 months then as needed monthly in accordance with standard of care. Patients receiving bromfenac self-administered 1 drop twice a day for 12 months. Patients were followed for 12 months. There were no safety concerns with the combination therapy. No statistically significant differences were identified in Early Treatment Diabetic Retinopathy Study best-corrected visual acuity or the number of injections required. However, the mean 12-month change in central macular thickness in the combination group was -81.56 μm while in the ranibizumab group alone the change was -42.50 μm (P = 0.03). The proportion of eyes experiencing a decrease in CMT of 50 μm or more was also significantly higher in those receiving combination therapy (P = 0.046). This pilot study is the first to prospectively identify a biologic signal that may indicate combination therapy with an easily administered well-tolerated eyedrop and ranibizumab is efficacious for the treatment of neovascular age-related macular degeneration. Further studies are warranted to validate this finding.
Sigma-Aldrich
Bromfenac sodium, ≥98% (HPLC)