Merck
  • A monobromobimane-based assay to measure the pharmacokinetic profile of reactive sulphide species in blood.
A monobromobimane-based assay to measure the pharmacokinetic profile of reactive sulphide species in blood.
British journal of pharmacology (2010-07-02)
Edward A Wintner, Thomas L Deckwerth, William Langston, Asa Bengtsson, Dina Leviten, Paul Hill, Michael A Insko, Ronald Dumpit, Emily VandenEkart, Christopher F Toombs, Csaba Szabo
ABSTRACT
Hydrogen sulphide (H(2)S) is a labile, endogenous metabolite of cysteine, with multiple biological roles. The development of sulphide-based therapies for human diseases will benefit from a reliable method of quantifying H(2)S in blood and tissues. Concentrations of reactive sulphide in saline and freshly drawn whole blood were quantified by reaction with the thio-specific derivatization agent monobromobimane, followed by reversed-phase fluorescence HPLC and/or mass spectrometry. In pharmacokinetic studies, male rats were exposed either to intravenous infusions of sodium sulphide or to H(2)S gas inhalation, and levels of available blood sulphide were measured. Levels of dissolved H(2)S/HS(-) were concomitantly measured using an amperometric sensor. Monobromobimane was found to rapidly and quantitatively derivatize sulphide in saline or whole blood to yield the stable small molecule sulphide dibimane. Extraction and quantification of this bis-bimane derivative were validated via reversed-phase HPLC separation coupled to fluorescence detection, and also by mass spectrometry. Baseline levels of sulphide in blood were in the range of 0.4-0.9 microM. Intravenous administration of sodium sulphide solution (2-20 mg x kg(-1) x h(-1)) or inhalation of H(2)S gas (50-400 ppm) elevated reactive sulphide in blood in a dose-dependent manner. Each 1 mg x kg(-1) x h(-1) of sodium sulphide infusion into rats was found to be pharmacokinetically equivalent to approximately 30 ppm of H(2)S gas inhalation. The monobromobimane derivatization method is a sensitive and reliable means to measure reactive sulphide species in whole blood. Using this method, we have established a bioequivalence between infused sodium sulphide and inhaled H(2)S gas.
Sigma-Aldrich
Bromobimane, ≥97% (HPLC)
Sigma-Aldrich
Bromobimane, suitable for fluorescence, BioReagent, ≥95% (HPCE)