Merck
  • Differential metabolomics for quantitative assessment of oxidative stress with strenuous exercise and nutritional intervention: thiol-specific regulation of cellular metabolism with N-acetyl-L-cysteine pretreatment.
Differential metabolomics for quantitative assessment of oxidative stress with strenuous exercise and nutritional intervention: thiol-specific regulation of cellular metabolism with N-acetyl-L-cysteine pretreatment.
Analytical chemistry (2010-03-03)
Richard Lee, Daniel West, Stuart M Phillips, Philip Britz-McKibbin
ABSTRACT
Despite several decades of active research, the success of large-scale clinical trials involving antioxidants remains equivocal given the complex biological interactions of reactive oxygen/nitrogen species in human health. Herein, we outline a differential metabolomics strategy by capillary electrophoresis-electrospray ionization-mass spectrometry (CE-ESI-MS) to assess the efficacy of nutritional intervention to attenuate oxidative stress induced by strenuous exercise. A healthy volunteer was recruited to perform a submaximal prolonged ergometer cycling trial until volitional exhaustion with frequent blood collection over a 6 h time interval, which included pre-, during, and postexercise periods while at rest. A follow-up study was subsequently performed by the same subject after high-dose oral intake of N-acetyl-L-cysteine (NAC) prior to performing the same exercise protocol under standardized conditions. Time-dependent changes in global metabolism of filtered red blood cell lysates by CE-ESI-MS were measured to reveal a significant attenuation of cellular oxidation associated with high-dose oral NAC intake relative to a control. Untargeted metabolite profiling allowed for the identification and quantification of several putative early- and late-stage biomarkers that reflected oxidative stress inhibition due to nutritional intervention, including oxidized glutathione (GSSG), reduced glutathione (GSH), 3-methylhistidine (3-MeHis), L-carnitine (C0), O-acetyl-L-carnitine (C2), and creatine (Cre). Our work demonstrates the proof-of-principle that NAC pretreatment is effective at dampening acute episodes of oxidative stress by reversible perturbations in global metabolism that can provide deeper insight into the mechanisms of thiol-specific protein inhibition relevant to its successful translation as a prophylaxis in clinical medicine.
Sigma-Aldrich
3-Methyl-L-histidine