Merck
Effects of phenylalaninol on centrally induced gastric acid secretion.
Chemical & pharmaceutical bulletin (1992-11-01)
H Hashizume, T Miyamae, T Morikawa, M Hagiwara
ABSTRACT
The effects of phenylalaninol (D-isomer) on gastric acid secretion and gastric ulcer were studied in rats. The compound reduced the gastric acid secretion stimulated by intracisternal thyrotropin releasing hormone and intravenous 2-deoxy-D-glucose, but not that stimulated by subcutaneous carbachol or histamine. Phenylalaninol prevented stress- and indomethacin-induced gastric ulcers. We conclude that phenylalaninol inhibits ulcer formation mainly by central inhibition of gastric acid secretion.
Sigma-Aldrich
(S)-(−)-2-Amino-3-phenyl-1-propanol, 98%, optical purity ee: 99% (HPLC)