Merck
  • Synthesis and biological profile of the enantiomers of [4-(4-amino-6,7-dimethoxyquinazolin-2-yl)-cis-octahydroquinoxalin- 1-yl]furan-2-ylmethanone (cyclazosin), a potent competitive alpha 1B- adrenoceptor antagonist.
Synthesis and biological profile of the enantiomers of [4-(4-amino-6,7-dimethoxyquinazolin-2-yl)-cis-octahydroquinoxalin- 1-yl]furan-2-ylmethanone (cyclazosin), a potent competitive alpha 1B- adrenoceptor antagonist.
Journal of medicinal chemistry (1996-11-08)
D Giardinà, M Crucianelli, R Romanelli, A Leonardi, E Poggesi, C Melchiorre
ABSTRACT
The enantiomers of [4-(4-amino-6, 7-dimethoxyquinazolin-2-yl)-cis-octahydroquinoxalin-1-yl]-fu ran- 2-ylmethanone (cyclazosin, 1) were synthesized from the chiral furan-2-yl(cis-octahydroquinoxalin-1-yl)methanone [(+)-2 and (-)-2], which were obtained by resolution of the racemic amine with (S)-(+)- and (R)-(-)-mandelic acid. The binding profile of the enantiomers of 1 was assessed at alpha 1-, alpha 2-, D2, and 5-HT1A receptors as well as at native alpha 1A- and alpha 1B- and cloned alpha 1a-, alpha 1b-, and alpha 1d-adrenoceptor subtypes in comparison with prazosin, spiperone, and AH11110A. (+)-1 displayed a 40-90-fold selectivity for the alpha 1B(alpha 1b)-adrenoceptor relative to alpha 1A(alpha 1a) and alpha 1d subtypes. A significant enantioselectivity was observed at the alpha 1A(alpha 1a)-adrenoceptor and particularly at alpha 1d-adrenoceptors since (-)-1 was 11-14- and 47-fold, respectively, more potent than (+)-1. Furthermore the enantiomer (+)-1 displayed selectivities of 1100-, 19000-, and 12000-fold in binding to alpha 1b-adrenoceptors relative to alpha 2-adrenoceptors and 5-HT1A and D2 receptors. These results indicate that (+)-1, [(+)-cyclazosin] is the most potent and selective ligand for the alpha 1B-adrenoceptor subtype so far described and may be a valuable tool in the characterization of alpha 1-adrenoceptor subtypes.
Sigma-Aldrich
4-Amino-2-chloro-6,7-dimethoxyquinazoline, 95%
Sigma-Aldrich
Cyclazosin hydrochloride, solid