Merck
  • Effects of Influenza Vaccine on the Immune Responses to SARS-CoV-2 Vaccination.
Effects of Influenza Vaccine on the Immune Responses to SARS-CoV-2 Vaccination.
Vaccines (2024-04-27)
A Riccomi, C M Trombetta, M Dorrucci, D Di Placido, N Sanarico, F Farchi, R Giuseppetti, U Villano, C Marcantonio, S Marchi, A Ciaramella, P Pezzotti, E Montomoli, C Valdarchi, A R Ciccaglione, S Vendetti
ABSTRACT
A number of studies have suggested that influenza vaccination can provide protection against COVID-19, but the underlying mechanisms that could explain this association are still unclear. In this study, the effect of the 2021/2022 seasonal influenza vaccination on the immune response to the booster dose of anti-SARS-CoV-2 vaccination was evaluated in a cohort of healthy individuals. A total of 113 participants were enrolled, 74 of whom had no prior COVID-19 diagnosis or significant comorbidities were considered for the analysis. Participants received the anti-influenza tetravalent vaccine and the booster dose of the anti-SARS-CoV-2 vaccine or the anti-SARS-CoV-2 vaccine alone. Blood was collected before and 4 weeks after each vaccination and 12 weeks after SARS-CoV-2 vaccination and analyzed for anti-flu and anti-spike-specific antibody titers and for in vitro influenza and SARS-CoV-2 neutralization capacity. Results indicated an increased reactivity in subjects who received both influenza and SARS-CoV-2 vaccinations compared to those who received only the SARS-CoV-2 vaccine, with sustained anti-spike antibody titers up to 12 weeks post-vaccination. Immune response to the influenza vaccine was evaluated, and individuals were stratified as high or low responders. High responders showed increased antibody titers against the SARS-CoV-2 vaccine both after 4 and 12 weeks post-vaccination. Conversely, individuals classified as low responders were less responsive to the SARS-CoV-2 vaccine. These data indicate that both external stimuli, such as influenza vaccination, and the host's intrinsic ability to respond to stimuli play a role in the response to the vaccine.
Sigma-Aldrich
Cholera filtrate, lyophilized powder