Merck
  • The prognostic impact of subclonal IDH1 mutation in grade 2-4 astrocytomas.
The prognostic impact of subclonal IDH1 mutation in grade 2-4 astrocytomas.
Neuro-oncology advances (2023-06-16)
Meenakshi Vij, Raquel T Yokoda, Omid Rashidipour, Ivy Tran, Varshini Vasudevaraja, Matija Snuderl, Raymund L Yong, William S Cobb, Melissa Umphlett, Jamie M Walker, Nadejda M Tsankova, Timothy E Richardson
ABSTRACT
Isocitrate dehydrogenase (IDH) mutations are thought to represent an early oncogenic event in glioma evolution, found with high penetrance across tumor cells; however, in rare cases, IDH mutation may exist only in a small subset of the total tumor cells (subclonal IDH mutation). We present 2 institutional cases with subclonal IDH1 R132H mutation. In addition, 2 large publicly available cohorts of IDH-mutant astrocytomas were mined for cases harboring subclonal IDH mutations (defined as tumor cell fraction with IDH mutation ≤0.67) and the clinical and molecular features of these subclonal cases were compared to clonal IDH-mutant astrocytomas. Immunohistochemistry (IHC) performed on 2 institutional World Health Organization grade 4 IDH-mutant astrocytomas revealed only a minority of tumor cells in each case with IDH1 R132H mutant protein, and next-generation sequencing (NGS) revealed remarkably low IDH1 variant allele frequencies compared to other pathogenic mutations, including TP53 and/or ATRX. DNA methylation classified the first tumor as high-grade IDH-mutant astrocytoma with high confidence (0.98 scores). In the publicly available datasets, subclonal IDH mutation was present in 3.9% of IDH-mutant astrocytomas (18/466 tumors). Compared to clonal IDH-mutant astrocytomas (n = 156), subclonal cases demonstrated worse overall survival in grades 3 (P = .0106) and 4 (P = .0184). While rare, subclonal IDH1 mutations are present in a subset of IDH-mutant astrocytomas of all grades, which may lead to a mismatch between IHC results and genetic/epigenetic classification. These findings suggest a possible prognostic role of IDH mutation subclonality, and highlight the potential clinical utility of quantitative IDH1 mutation evaluation by IHC and NGS.
Sigma-Aldrich
Anti-APC11 Antibody, clone CRQ1.1, clone CRQ1.1, from mouse