Merck
Transferrin and transferrin receptors update.
Free radical biology & medicine (2018-07-04)
Hiroshi Kawabata
ABSTRACT
In vertebrates, transferrin (Tf) safely delivers iron through circulation to cells. Tf-bound iron is incorporated through Tf receptor (TfR) 1-mediated endocytosis. TfR1 can mediate cellular uptake of both Tf and H-ferritin, an iron storage protein. New World arenaviruses, which cause hemorrhagic fever, and Plasmodium vivax use TfR1 for entry into host cells. Human TfR2, another receptor for Tf, is predominantly expressed in hepatocytes and erythroid precursors, and holo-Tf dramatically upregulates its expression. TfR2 forms a complex with hemochromatosis protein, HFE, and serves as a component of the iron sensing machinery in hepatocytes. Defects in TfR2 cause systemic iron overload, hemochromatosis, through down-regulation of hepcidin. In erythroid cells, TfR2 forms a complex with the erythropoietin receptor and regulates erythropoiesis. TfR2 facilitates iron transport from lysosomes to mitochondria in erythroblasts and dopaminergic neurons. Administration of apo-Tf, which scavenges free iron, has been explored for various clinical conditions including atransferrinemia, iron overload, and tissue ischemia. Apo-Tf has also been shown to ameliorate anemia in animal models of β-thalassemia. In this review, I provide an update and summary on our knowledge of mammalian Tf and its receptors.
Sigma-Aldrich
apo-Transferrin human, γ-irradiated, powder, BioXtra, suitable for cell culture, ≥98%