Merck
Formation and oxidation state of CeO(2-x) nanotubes.
Journal of the American Chemical Society (2005-09-15)
Wei-Qiang Han, Lijun Wu, Yimei Zhu
ABSTRACT
Well-crystalline CeO(2-x) nanotubes are synthesized via a mild hydrothermal reaction route using cerium nitrate and ammonium hydroxide as reactants. The CeO(2-x) nanotubes have the same structure as the bulk CeO2 but larger lattice parameter. The measurement of the ratio of M5/M4 edge shows the valence reduction of cerium ions for the CeO(2-x) nanotubes.
Sigma-Aldrich
Cerium(III) nitrate hexahydrate, 99% trace metals basis