Merck
  • Spatially resolved deconvolution of the fibrotic niche in lung fibrosis.
Spatially resolved deconvolution of the fibrotic niche in lung fibrosis.
Cell reports (2022-08-18)
Michael Eyres, Joseph A Bell, Elizabeth R Davies, Aurelie Fabre, Aiman Alzetani, Sanjay Jogai, Ben G Marshall, David A Johnston, Zijian Xu, Sophie V Fletcher, Yihua Wang, Gayle Marshall, Donna E Davies, Emily Offer, Mark G Jones
ABSTRACT
A defining pathological feature of human lung fibrosis is localized tissue heterogeneity, which challenges the interpretation of transcriptomic studies that typically lose spatial information. Here we investigate spatial gene expression in diagnostic tissue using digital profiling technology. We identify distinct, region-specific gene expression signatures as well as shared gene signatures. By integration with single-cell data, we spatially map the cellular composition within and distant from the fibrotic niche, demonstrating discrete changes in homeostatic and pathologic cell populations even in morphologically preserved lung, while through ligand-receptor analysis, we investigate cellular cross-talk within the fibrotic niche. We confirm findings through bioinformatic, tissue, and in vitro analyses, identifying that loss of NFKB inhibitor zeta in alveolar epithelial cells dysregulates the TGFβ/IL-6 signaling axis, which may impair homeostatic responses to environmental stress. Thus, spatially resolved deconvolution advances understanding of cell composition and microenvironment in human lung fibrogenesis.
Sigma-Aldrich
Anti-NFKBIZ antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.