Merck
  • Immunization with synthetic SARS-CoV-2 S glycoprotein virus-like particles protects macaques from infection.
Immunization with synthetic SARS-CoV-2 S glycoprotein virus-like particles protects macaques from infection.
Cell reports. Medicine (2022-03-03)
Guidenn Sulbaran, Pauline Maisonnasse, Axelle Amen, Gregory Effantin, Delphine Guilligay, Nathalie Dereuddre-Bosquet, Judith A Burger, Meliawati Poniman, Marloes Grobben, Marlyse Buisson, Sebastian Dergan Dylon, Thibaut Naninck, Julien Lemaître, Wesley Gros, Anne-Sophie Gallouët, Romain Marlin, Camille Bouillier, Vanessa Contreras, Francis Relouzat, Daphna Fenel, Michel Thepaut, Isabelle Bally, Nicole Thielens, Franck Fieschi, Guy Schoehn, Sylvie van der Werf, Marit J van Gils, Rogier W Sanders, Pascal Poignard, Roger Le Grand, Winfried Weissenhorn
ABSTRACT
The severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) pandemic has caused an ongoing global health crisis. Here, we present as a vaccine candidate synthetic SARS-CoV-2 spike (S) glycoprotein-coated lipid vesicles that resemble virus-like particles. Soluble S glycoprotein trimer stabilization by formaldehyde cross-linking introduces two major inter-protomer cross-links that keep all receptor-binding domains in the "down" conformation. Immunization of cynomolgus macaques with S coated onto lipid vesicles (S-LVs) induces high antibody titers with potent neutralizing activity against the vaccine strain, Alpha, Beta, and Gamma variants as well as T helper (Th)1 CD4+-biased T cell responses. Although anti-receptor-binding domain (RBD)-specific antibody responses are initially predominant, the third immunization boosts significant non-RBD antibody titers. Challenging vaccinated animals with SARS-CoV-2 shows a complete protection through sterilizing immunity, which correlates with the presence of nasopharyngeal anti-S immunoglobulin G (IgG) and IgA titers. Thus, the S-LV approach is an efficient and safe vaccine candidate based on a proven classical approach for further development and clinical testing.