Merck
ICP-MS trace element analysis in serum and whole blood.
PloS one (2020-05-21)
Nico Laur, Ralf Kinscherf, Karolina Pomytkin, Lars Kaiser, Otto Knes, Hans-Peter Deigner
ABSTRACT
Trace elements and minerals are compounds that are essential for the support of a variety of biological functions and play an important role in the formation of and the defense against oxidative stress. Here we describe a technique, allowing sequential detection of the trace elements (K, Zn, Se, Cu, Mn, Fe, Mg) in serum and whole blood by an ICP-MS method using single work-up, which is a simple, quick and robust method for the sequential measurement and quantification of the trace elements Sodium (Na), Potassium (K), Calcium (Ca), Zinc (Zn), Selenium (Se), Copper (Cu), Iron (Fe), Manganese (Mn) and Magnesium (Mg) in whole blood as well as Copper (Cu), Selenium (Se), Zinc (Zn), Iron (Fe), Magnesium (Mg), Manganese (Mn), Chromium (Cr), Nickel (Ni), Gold (Au) and Lithium (Li) in human serum. For analysis, only 100 μl of serum or whole blood is sufficient, which make this method suitable for detecting trace element deficiency or excess in newborns and infants. All samples were processed and analyzed by ICP-MS (Agilent Technologies). The accuracy, precision, linearity and the limit of quantification (LOQ), Limit of Blank (LOB) and the limit of detection (LOD) of the method were assessed. Recovery rates were between 80-130% for most of the analyzed elements; repeatabilities (Cv %) calculated were below 15% for most of the measured elements. The validity of the proposed methodology was assessed by analyzing a certified human serum and whole blood material with known concentrations for all elements; the method described is ready for routine use in biomonitoring studies.
Sigma-Aldrich
ITS Liquid Media Supplement (100×), liquid, sterile-filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
SITE Liquid Media Supplement (100×), liquid, sterile-filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.