Merck
  • Identification of a series of hair-cell MET channel blockers that protect against aminoglycoside-induced ototoxicity.
Identification of a series of hair-cell MET channel blockers that protect against aminoglycoside-induced ototoxicity.
JCI insight (2021-03-19)
Emma J Kenyon, Nerissa K Kirkwood, Siân R Kitcher, Richard J Goodyear, Marco Derudas, Daire M Cantillon, Sarah Baxendale, Antonio de la Vega de León, Virginia N Mahieu, Richard T Osgood, Charlotte Donald Wilson, James C Bull, Simon J Waddell, Tanya T Whitfield, Simon E Ward, Corné J Kros, Guy P Richardson
ABSTRACT
To identify small molecules that shield mammalian sensory hair cells from the ototoxic side effects of aminoglycoside antibiotics, 10,240 compounds were initially screened in zebrafish larvae, selecting for those that protected lateral-line hair cells against neomycin and gentamicin. When the 64 hits from this screen were retested in mouse cochlear cultures, 8 protected outer hair cells (OHCs) from gentamicin in vitro without causing hair-bundle damage. These 8 hits shared structural features and blocked, to varying degrees, the OHC's mechano-electrical transducer (MET) channel, a route of aminoglycoside entry into hair cells. Further characterization of one of the strongest MET channel blockers, UoS-7692, revealed it additionally protected against kanamycin and tobramycin and did not abrogate the bactericidal activity of gentamicin. UoS-7692 behaved, like the aminoglycosides, as a permeant blocker of the MET channel; significantly reduced gentamicin-Texas red loading into OHCs; and preserved lateral-line function in neomycin-treated zebrafish. Transtympanic injection of UoS-7692 protected mouse OHCs from furosemide/kanamycin exposure in vivo and partially preserved hearing. The results confirmed the hair-cell MET channel as a viable target for the identification of compounds that protect the cochlea from aminoglycosides and provide a series of hit compounds that will inform the design of future otoprotectants.
Sigma-Aldrich
Tobramycin sulfate salt, aminoglycoside antibiotic
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, powder, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Gentamicin sulfate salt, potency: ≥590 I.U. Gentamicin base per mg