Merck
  • Sex dependent glial-specific changes in the chromatin accessibility landscape in late-onset Alzheimer's disease brains.
Sex dependent glial-specific changes in the chromatin accessibility landscape in late-onset Alzheimer's disease brains.
Molecular neurodegeneration (2021-08-26)
Julio Barrera, Lingyun Song, Julia E Gamache, Melanie E Garrett, Alexias Safi, Young Yun, Ivana Premasinghe, Daniel Sprague, Danielle Chipman, Jeffrey Li, Hélène Fradin, Karen Soldano, Raluca Gordân, Allison E Ashley-Koch, Gregory E Crawford, Ornit Chiba-Falek
ABSTRACT
In the post-GWAS era, there is an unmet need to decode the underpinning genetic etiologies of late-onset Alzheimer's disease (LOAD) and translate the associations to causation. We conducted ATAC-seq profiling using NeuN sorted-nuclei from 40 frozen brain tissues to determine LOAD-specific changes in chromatin accessibility landscape in a cell-type specific manner. We identified 211 LOAD-specific differential chromatin accessibility sites in neuronal-nuclei, four of which overlapped with LOAD-GWAS regions (±100 kb of SNP). While the non-neuronal nuclei did not show LOAD-specific differences, stratification by sex identified 842 LOAD-specific chromatin accessibility sites in females. Seven of these sex-dependent sites in the non-neuronal samples overlapped LOAD-GWAS regions including APOE. LOAD loci were functionally validated using single-nuclei RNA-seq datasets. Using brain sorted-nuclei enabled the identification of sex-dependent cell type-specific LOAD alterations in chromatin structure. These findings enhance the interpretation of LOAD-GWAS discoveries, provide potential pathomechanisms, and suggest novel LOAD-loci.
Sigma-Aldrich
Milli-Mark® Anti-NeuN-PE Antibody, clone A60, clone A60, Milli-Mark®, from mouse