Merck
  • Microcystin-leucine-arginine induces liver fibrosis by activating the Hedgehog pathway in hepatic stellate cells.
Microcystin-leucine-arginine induces liver fibrosis by activating the Hedgehog pathway in hepatic stellate cells.
Biochemical and biophysical research communications (2020-09-30)
Shen Gu, Minghao Yan, Cong Wang, Xiannan Meng, Zou Xiang, Yudong Qiu, Xiaodong Han
ABSTRACT
Microcystin-leucine-arginine (MC-LR), produced by cyanobacteria, accumulates in the liver through blood circulation. We investigated the impact of MC-LR on liver fibrosis. Mice received a daily injection of MC-LR at various concentrations for 14 consecutive days aa and then mouse liver was obtained for histopathological and immunoblot analysis. Next, a human hepatic stellate cell line (LX-2) was treated with MC-LR at various concentrations followed by measurement of cell viability, cell cycle and relevant protein expression levels. Our data confirmed the induction of mouse liver fibrosis after exposure to MC-LR at 15 μg/kg and 30 μg/kg. Furthermore, we demonstrated that LX-2 cells could uptake MC-LR, resulting in cell proliferation and differentiation through impacting the Hedgehog signaling after the treatment of MC-LR at 50 nM. Our data supported that MC-LR could induce liver fibrosis by modulating the expression of the transcription factor Gli2 in the Hedgehog signaling in hepatic stellate cells.
Sigma-Aldrich
GANT58, ≥98% (HPLC)