Merck
  • Internal Microscopic Diagnosis of Accelerated Aging of Proton Exchange Membrane Water Electrolysis Cell Stack.
Internal Microscopic Diagnosis of Accelerated Aging of Proton Exchange Membrane Water Electrolysis Cell Stack.
Micromachines (2020-12-10)
Chi-Yuan Lee, Chia-Hung Chen, Guo-Bin Jung, Shih-Chun Li, Yi-Zhen Zeng
ABSTRACT
The hydrogen production reaction of the proton exchange membrane (PEM) water electrolysis cell stack is the reverse reaction of the fuel cell, but the water electrolysis operation requires high pressure, and the high pressure decomposes hydrogen molecules, thus aging or causing failure in the water electrolysis cell stack. In addition, there are five important physical parameters (current, voltage, flow, pressure and temperature) inside the water electrolysis cell stack, which can change the performance and shorten the life of the cell stack. However, the present techniques obtain data only by external simulation or single measurement; they cannot collect the internal real data in operation instantly and accurately. This study discusses the causes for aging or failure, and develops an internal real-time microscopic diagnosis tool for accelerated aging of the PEM water electrolysis cell stack. A flexible integrated (current, voltage, flow, pressure and temperature) microsensor applicable to the inside (high voltage and electrochemical environment) of the PEM water electrolysis cell stack is developed by using micro-electro-mechanical systems (MEMS) technology; it is embedded in the PEM water electrolysis cell stack for microscopic diagnosis of accelerated aging, and 100-h durability and reliability tests are performed. The distribution of important physical parameters inside the PEM water electrolysis cell stack can be measured instantly and accurately, so as to adjust it to the optimal operating conditions, and the local aging and failure problems are discussed.
Sigma-Aldrich
PD 146176, ≥98% (HPLC), solid