Merck
  • Chemical inhibition of the histone acetyltransferase activity in Arabidopsis thaliana.
Chemical inhibition of the histone acetyltransferase activity in Arabidopsis thaliana.
Biochemical and biophysical research communications (2016-12-21)
Felipe Aquea, Tania Timmermann, Ariel Herrera-Vásquez
ABSTRACT
Chemical inhibition of chromatin regulators provides an effective approach to investigate the roles of chromatin modifications in plant and animals. In this work, chemical inhibition of the Arabidopsis histone acetyltransferase activity by γ-butyrolactone (MB-3), the inhibitor of the catalytic activity of mammalian GENERAL CONTROL NON-REPRESSIBLE 5 (GCN5) is evaluated. Arabidopsis seedlings were germinated in LS medium supplemented with different concentrations of MB-3, and inhibition in the root length and yellowed leaves were observed. The yellowed leaves phenotype of the plants grown in 100 μM of MB-3 was reverted when plants were additionally treated with 1 μM of TSA, a histone deacetylase inhibitor. Using an immunoblot assay with specific antibodies revealed a reduction of H3K14 acetylation levels at 3 and 24 h post-treatment. At 24 h post-treatment a reduction of H3K9 acetylation levels was observed. Targets of GCN5 related to stress were downregulated at 3 h post-treatment but no change was observed in target genes related to developmental transition. Our results indicate that MB-3 is a chemical inhibitor of the histone acetyltransferase in Arabidopsis and suggest that this inhibitor could function in other plants species.
Sigma-Aldrich
MB-3, ≥95% (HPLC)