Merck
Rational design of novel glycomimetics: inhibitors of concanavalin A.
Bioorganic & medicinal chemistry letters (2008-11-08)
Karen T Welch, Trent A Turner, Callie E Preast
ABSTRACT
A virtual screening approach was used to identify new glycomimetics. The National Cancer Institute Diversity Set was docked into the carbohydrate binding site of the lectin concanavalin A (ConA). The resulting poses were analyzed and 19 molecules were tested for inhibition with an enzyme-linked lectin assay (ELLA). Eight of the 19 molecules inhibited ConA-carbohydrate binding. The two most potent inhibitors have IC(50) values that are an order of magnitude smaller than the monosaccharide methyl alpha-D-mannopyranoside.
Product Description
Millipore
Methyl α-D-mannopyranoside, suitable for microbiology, ≥99.0%, enables differentiation between species of Listeria
Sigma-Aldrich
Methyl α-D-mannopyranoside, ≥99.0% (HPLC)