Merck
Visualizing the Itch-Sensing Skin Arbors.
The Journal of investigative dermatology (2020-10-23)
Yanyan Xing, Haley R Steele, Henry B Hilley, Yuyan Zhu, Katy Lawson, Taylor Niehoff, Liang Han, Yanyan Xing, Haley R Steele, Henry B Hilley, Yuyan Zhu, Katy Lawson, Taylor Niehoff, Liang Han
ABSTRACT
Diverse sensory neurons exhibit distinct neuronal morphologies with a variety of axon terminal arborizations subserving their functions. Because of its clinical significance, the molecular and cellular mechanisms of itch are being intensely studied. However, a complete analysis of itch-sensing terminal arborization is missing. Using an MrgprC11CreERT2 transgenic mouse line, we labeled a small subset of itch-sensing neurons that express multiple itch-related molecules including MrgprA3, MrgprC11, histamine receptor H1, IL-31 receptor, 5-hydroxytryptamine receptor 1F, natriuretic precursor peptide B, and neuromedin B. By combining sparse genetic labeling and whole-mount placental alkaline phosphatase histochemistry, we found that itch-sensing skin arbors exhibit free endings with extensive axonal branching in the superficial epidermis and large receptive fields. These results revealed the unique morphological characteristics of itch-sensing neurons and provide intriguing insights into the basic mechanisms of itch transmission.
Sigma-Aldrich
Anti-Protein Gene Product 9.5 Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Substance P Antibody, pain, clone NC1, culture supernatant, clone NC1, Chemicon®