Merck
  • Low molecular weight polyethyleneimine conjugated guar gum for targeted gene delivery to triple negative breast cancer.
Low molecular weight polyethyleneimine conjugated guar gum for targeted gene delivery to triple negative breast cancer.
International journal of biological macromolecules (2020-06-20)
Piyali Jana, Santanu Ghosh, Kishor Sarkar
ABSTRACT
The study emphasized on the development of an efficient, receptor-targeted non-viral gene delivery vehicle for gene therapy of triple negative breast cancer (TNBC). Here, naturally abundant guar gum based non-viral carrier was developed through conjugating by low molecular weight polyethylenimine (LPEI) (GNP) using napthalic anhydride coupling agent and characterized them by FT-IR, 1H NMR, XRD and UV spectrophotometer. The carrier was found to be cytocompatible as revealed by MTT assay against MDA-MB-231 and HeLa cell lines and excellent blood compatibility till the concentration of 200 μg/ml. In addition to these, the carrier exhibited excellent gene binding capability and formed spherical shaped complexes. The carrier showed very high in vitro transfection efficiency in TNBC cell (MDA-MB-231) compared to lipofectamine 2000 (LF2k) which could be justified by its high buffering capacity. Therefore, GNP may be an attractive non-viral gene carrier for gene therapy of TNBC in future.
Sigma-Aldrich
4-Bromo-1,8-naphthalic anhydride, 95%