Merck
  • Signal optimized rough silver nanoparticle for rapid SERS sensing of pesticide residues in tea.
Signal optimized rough silver nanoparticle for rapid SERS sensing of pesticide residues in tea.
Food chemistry (2020-08-18)
Md Mehedi Hassan, Muhammad Zareef, Tianhui Jiao, Shuangshuang Liu, Yi Xu, Annavaram Viswadevarayalu, Huanhuan Li, Quansheng Chen
ABSTRACT
Trace detection of toxic chemicals in foodstuffs is of great concern in recent years. Surface-enhanced Raman scattering (SERS) has drawn significant attention in the monitoring of food safety due to its high sensitivity. This study synthesized signal optimized flower-like silver nanoparticle-(AgNP) with EF at 25 °C of 1.39 × 106 to extend the SERS application for pesticide sensing in foodstuffs. The synthesized AgNP was deployed as SERS based sensing platform to detect methomyl, acetamiprid-(AC) and 2,4-dichlorophenoxyacetic acid-(2,4-D) residue levels in green tea via solid-phase extraction. A linear correlation was twigged between the SERS signal and the concentration for methomyl, AC and 2,4-D with regression coefficient of 0.9974, 0.9956 and 0.9982 and limit of detection of 5.58 × 10-4, 1.88 × 10-4 and 4.72 × 10-3 µg/mL, respectively; the RSD value < 5% was recorded for accuracy and precision analysis suggesting that proposed method could be deployed for the monitoring of methomyl, AC and 2,4-D residue levels in green tea.
Product Description
Supelco
Supelclean ENVI-Carb SPE Tube, bed wt. 500 mg, volume 6 mL, pk of 30