Merck
  • SKF83959, an agonist of phosphatidylinositol-linked dopamine receptors, prevents renewal of extinguished conditioned fear and facilitates extinction.
SKF83959, an agonist of phosphatidylinositol-linked dopamine receptors, prevents renewal of extinguished conditioned fear and facilitates extinction.
Brain research (2020-09-28)
Fu-Feng Chen, Can-Ming Wang, Hong-Sheng Chen, Ji Wang, Qian-Qian Han, Yu Cao, Tian-Tian Shen, Yuan-Jian Yang, Zhuang-Li Hu, Fang Wang, Jian-Guo Chen, Peng-Fei Wu
ABSTRACT
Fear-related anxiety disorders, such as social phobia and post-traumatic stress disorder, are partly explained by an uncontrollable state of fear. An emerging literature suggests dopamine receptor-1 (D1 receptor) in the amygdala is involved in the regulation of fear memory. An early study has reported that amygdaloid D1 receptor (D1R) is not coupled to the classic cAMP-dependent signal transduction. Here, we investigated whether SKF83959, a typical D1R agonist that mainly activates a D1-like receptor-dependent phosphatidylinositol (PI) signal pathway, facilitates fear extinction and reduces the return of extinguished fear. Interestingly, long-term loss of fearful memories can be induced through a combination of SKF83959 (1 mg/kg/day, i.p., once daily for one week) pharmacotherapy and extinction training. Furthermore, sub-chronic administration of SKF83959 after fear conditioning reduced fear renewal and reinstatement in the mice. We found that the activation D1R and PI signaling in the amygdala was responsible for the effect of SKF83959 on fear extinction. Additionally, SKF83959 significantly promoted the elevation of brain-derived neurotrophic factor (BDNF) expression, possibly by the cAMP response element binding protein (CREB) -directed gene transcription. Given the beneficial effects on extinction, SKF83959 may emerge as a candidate pharmacological approach for improving cognitive-behavioral therapy on fear-related anxiety disorders.
Sigma-Aldrich
L-741,626, ≥98% (HPLC)