- Methylene-bridged bis(benzimidazolium) salt as a highly efficient catalyst for the benzoin reaction in aqueous media.
Methylene-bridged bis(benzimidazolium) salt as a highly efficient catalyst for the benzoin reaction in aqueous media.
Organic & biomolecular chemistry (2008-02-23)
Ken-ichi Iwamoto, Hitomi Kimura, Masaaki Oike, Masayuki Sato
PMID18292884
ABSTRACT
Benzoin reactions are catalyzed effectively by a methylene-bridged bis(benzimidazolium) salt to yield alpha-hydroxy ketones, and the reactions proceed in water as the aqueous medium under mild conditions.