Merck
  • Extending the family of Zn-based MOFs: synthetic approaches to chiral framework structures and MOFs with large pores and channels.
Extending the family of Zn-based MOFs: synthetic approaches to chiral framework structures and MOFs with large pores and channels.
Chemical communications (Cambridge, England) (2012-03-07)
Nianyong Zhu, Gerard Tobin, Wolfgang Schmitt
ABSTRACT
Tri- and pentanuclear, kinetically stable SBUs were exploited for the preparation of the novel MOFs [Zn(3)(BTEB)(2)(DMF)(2)] and (Me(2)NH(2))[Zn(5)(BTEB)(3)(μ(3)-OH)(2)(DMF)(2)]. The applied synthetic approach results in topologies that are stabilised by tritopic benzene-trisethynylbenzoic acid (BTEB) linkers giving rise to chiral frameworks with large pores or channels.
Sigma-Aldrich
1,3,5-Triscarboxyphenylethynylbenzene, ≥95%