Merck
  • Aberrant methylation and microRNA-target regulation are associated with downregulated NEURL1B: a diagnostic and prognostic target in colon cancer.
Aberrant methylation and microRNA-target regulation are associated with downregulated NEURL1B: a diagnostic and prognostic target in colon cancer.
Cancer cell international (2020-08-04)
Jiaxin Liu, Zhao Liu, Xiaozhi Zhang, Yanli Yan, Shuai Shao, Demao Yao, Tuotuo Gong
ABSTRACT
Aberrant methylation and miRNA-target-gene regulation function as important mechanisms for gene inactivation in colon carcinogenesis. Although a serious of molecular events (such as aberrant alterations of genomics and epigenetics) have been identified to be related to prognostic in colon cancer (CC) patients, beneficial biomarkers for early diagnosis and prognostic evaluation remain largely unknown. In our study, the role of NEURL1B, including gene expression analysis, methylation characteristic, miRNA-target regulation, diagnostic and prognostic significance, were evaculated using multiple bioinformatic tools based on TCGA database and clinical samples. Our data showed that NEURL1B was aberrantly downregulated in CC, regardless of the mRNA level or protein level. Moreover, ROC curve and multivariate Cox regression analysis demonstrated that NEURL1B was a diagnostic and independent prognostic facter for CC patients. Of interest, methylation of NEURL1B was also high and closely associated with poor survival in CC. In addition, multiple NEURL1B-target miRNAs were found to be overexpressed in CC tissues. Thus, our findings suggested that NEURL1B participated in the pathological processes of CC as a tumor suppressor gene. Double management, including DNA methylation modification and miRNA-target regulation, were considered to be related to the downregulation of NEURL1B. Importantly, there existing be an significant intersection between miRNAs-target pathways and NEURL1B-target pathways, suggesting that miR-17 and miR-27a might promote tumor cell malignant property by targeting NEURL1B degradation via the activation of PI3K/AKT signaling pathway. Taking together, the first investigation of NEURL1B in CC provide us a strong evidences that it might be served as a potential biomarkers for early diagnosis and prognostic evaluation in CC.