MilliporeSigma
Co-evaporated bulk heterojunction solar cells with >6.0% efficiency.
Advanced materials (Deerfield Beach, Fla.) (2012-04-20)
Guo Chen, Hisahiro Sasabe, Zhongqiang Wang, Xiao-Feng Wang, Ziruo Hong, Yang Yang, Junji Kido
ABSTRACT
High power conversion efficiencies of over 6.0% are achieved with a squaraine compound from co-deposited photovoltaic cells with a simple device structure, in which the efficiency is insensitive to blending ratios and thicknesses of photoactive layers. It demonstrates the huge potential of low molecular weight materials in photovoltaic cells via vacuum processes.