Merck
  • Elevated galanin receptor type 2 primarily contributes to mechanical hypersensitivity after median nerve injury.
Elevated galanin receptor type 2 primarily contributes to mechanical hypersensitivity after median nerve injury.
PloS one (2018-06-22)
Seu-Hwa Chen, June-Horng Lue, Yung-Jung Hsiao, Shu-Mei Lai, Hsin-Ying Wang, Chi-Te Lin, Ya-Chin Chen, Yi-Ju Tsai
ABSTRACT
In this study, we investigated temporal changes in galanin receptor type 2 (GalR2) expression in NF200-, galanin-, neuropeptide Y (NPY)-, and neuronal nitric oxide synthase (nNOS)-like immunoreactive (LI) dorsal root ganglion (DRG) neurons after median nerve chronic constriction injury (CCI), and the effects of GalR2 on c-Fos expression in the cuneate nucleus (CN). Double immunofluorescence labeling methods were used to appraise changes in GalR2 expression in NF200-LI, galanin-LI, NPY-LI, and nNOS-LI DRG neurons after CCI. The von Frey assay was used to assess the efficiency of intraplantar administration of saline, M871 (a GalR2 antagonist), or AR-M1896 (a GalR2 agonist) on neuropathic signs of rats with CCI. The effects of alterations in c-Fos expression were assessed in all treatments. The percentage of GalR2-LI neurons in lesioned DRGs increased and peaked at 1 week after CCI. We further detected that percentages of GalR2-LI neurons labeled for NF200, galanin, NPY, and nNOS significantly increased following CCI. Furthermore, M871 remarkably attenuated tactile allodynia, but the sensation was slightly aggravated by AR-M1896 after CCI. Consequentially, after electrical stimulation of the CCI-treated median nerve, the number of c-Fos-LI neurons in the cuneate nucleus (CN) was significantly reduced in the M871 group, whereas it increased in the AR-M1896 group. These results suggest that activation of GalR2, probably through NPY or nitric oxide, induces c-Fos expression in the CN and transmits mechanical allodynia sensations to the thalamus.
Sigma-Aldrich
M871 trifluoroacetate, ≥95% (HPLC)