Merck
  • Structural Peculiarities and Thermoelectric Study of Iron Indium Thiospinel.
Structural Peculiarities and Thermoelectric Study of Iron Indium Thiospinel.
Chemistry (Weinheim an der Bergstrasse, Germany) (2020-01-17)
Paweł Wyżga, Igor Veremchuk, Matej Bobnar, Primož Koželj, Steffen Klenner, Rainer Pöttgen, Andreas Leithe-Jasper, Roman Gumeniuk
ABSTRACT
The homogeneity range of ternary iron indium thiospinel at 873 K was investigated. A detailed study was focused on two distinct series (y=z): 1) a previously reported charge-balanced (In0.67+0.33y □0.33-0.33y )tetr [In2-z Fez ]oct S4 (A1-series; □ stands for vacancy; the abbreviations "tetr" and "oct" indicate atoms occupying tetrahedral 8a and octahedral 16d sites, respectively) and 2) a new charge-unbalanced (In0.67+y □0.33-y )tetr [In2-z Fez ]oct S4 (A2-series). Fe atoms were confirmed to exclusively occupy an octahedral position in both series. An unusual reduction of the unit cell parameter with increasing Fe content is explained by differences in the ionic radii between Fe and In, as well as by an additional electrostatic attraction originating from charge imbalance (latter only in A2-series). The studied compound is an n-type semiconductor, and its charge carrier concentration increases or decreases for larger Fe content within the A1- and A2-series, respectively. The thermal conductivity κtot is significantly reduced upon increasing vacancy concentration, whereas the change of power factor is insufficient to drastically improve the thermoelectric figure of merit.
Sigma-Aldrich
Sulfur, purum p.a., ≥99% (T)