Merck
  • Reaction rate of pyruvate and hydrogen peroxide: assessing antioxidant capacity of pyruvate under biological conditions.
Reaction rate of pyruvate and hydrogen peroxide: assessing antioxidant capacity of pyruvate under biological conditions.
Scientific reports (2019-12-22)
Victoria A Guarino, William M Oldham, Joseph Loscalzo, Ying-Yi Zhang
ABSTRACT
Pyruvate, a pivotal glucose metabolite, is an α-ketoacid that reacts with hydrogen peroxide (H2O2). Its pharmacological precursor, ethyl pyruvate, has shown anti-inflammatory/anti-tissue injury effects in various animal models of disease, but failed in a multicenter clinical trial. Since rodents, but not humans, can convert ethyl pyruvate to pyruvate in blood plasma, this additional source of extracellular pyruvate may have contributed to the discrepancy between the species. To examine this possibility, we investigated the kinetics of the reaction under biological conditions and determined the second order rate constant k as 2.360 ± 0.198 M-1 s-1. We then calculated the time required for H2O2 elimination by pyruvate. The results show that, with an average intracellular concentration of pyruvate (150 µM), elimination of 95% H2O2 at normal to pathological concentrations (0.01-50 µM) requires 141-185 min (2.4-3 hour). With 1,000 µM pyruvate, a concentration that can only exist extracellularly or in cell culture media, 95% elimination of H2O2 at 5-200 µM requires 21-25 min. We conclude that intracellular pyruvate, or other α-ketoacids, whose endogenous concentration is controlled by metabolism, have little role in H2O2 clearance. An increased extracellular concentration of pyruvate, however, does have remarkable peroxide scavenging effects, considering minimal peroxidase activity in this space.
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sodium acetate trihydrate, BioUltra, ≥99.5% (NT)