Merck
  • Combined proteomics/miRNomics of dendritic cell immunotherapy-treated glioblastoma patients as a screening for survival-associated factors.
Combined proteomics/miRNomics of dendritic cell immunotherapy-treated glioblastoma patients as a screening for survival-associated factors.
NPJ vaccines (2020-01-24)
Friedrich Erhart, Matthias Hackl, Hannes Hahne, Johanna Buchroithner, Chen Meng, Simone Klingenbrunner, René Reitermaier, Katrin Fischhuber, Susanna Skalicky, Walter Berger, Sabine Spiegl-Kreinecker, Daniela Lötsch, Gerda Ricken, Bernhard Kuster, Adelheid Wöhrer, Georg Widhalm, Johannes Hainfellner, Thomas Felzmann, Alexander M Dohnal, Christine Marosi, Carmen Visus
ABSTRACT
Glioblastoma is the most prevalent and aggressive brain cancer. With a median overall survival of ~15-20 months under standard therapy, novel treatment approaches are desperately needed. A recent phase II clinical trial with a personalized immunotherapy based on tumor lysate-charged dendritic cell (DC) vaccination, however, failed to prolong survival. Here, we investigated tumor tissue from trial patients to explore glioblastoma survival-related factors. We followed an innovative approach of combining mass spectrometry-based quantitative proteomics (n = 36) with microRNA sequencing plus RT-qPCR (n = 38). Protein quantification identified, e.g., huntingtin interacting protein 1 (HIP1), retinol-binding protein 1 (RBP1), ferritin heavy chain (FTH1) and focal adhesion kinase 2 (FAK2) as factor candidates correlated with a dismal prognosis. MicroRNA analysis identified miR-216b, miR-216a, miR-708 and let-7i as molecules potentially associated with favorable tissue characteristics as they were enriched in patients with a comparably longer survival. To illustrate the utility of integrated miRNomics and proteomics findings, focal adhesion was studied further as one example for a pathway of potential general interest. Taken together, we here mapped possible drivers of glioblastoma outcome under immunotherapy in one of the largest DC vaccination tissue analysis cohorts so far-demonstrating usefulness and feasibility of combined proteomics/miRNomics approaches. Future research should investigate agents that sensitize glioblastoma to (immuno)therapy-potentially building on insights generated here.
Sigma-Aldrich
1,2,4,5-Benzenetetramine tetrahydrochloride, ≥95%