Merck
  • Frequency of mononuclear diploid cardiomyocytes underlies natural variation in heart regeneration.
Frequency of mononuclear diploid cardiomyocytes underlies natural variation in heart regeneration.
Nature genetics (2017-08-08)
Michaela Patterson, Lindsey Barske, Ben Van Handel, Christoph D Rau, Peiheng Gan, Avneesh Sharma, Shan Parikh, Matt Denholtz, Ying Huang, Yukiko Yamaguchi, Hua Shen, Hooman Allayee, J Gage Crump, Thomas I Force, Ching-Ling Lien, Takako Makita, Aldons J Lusis, S Ram Kumar, Henry M Sucov
ABSTRACT
Adult mammalian cardiomyocyte regeneration after injury is thought to be minimal. Mononuclear diploid cardiomyocytes (MNDCMs), a relatively small subpopulation in the adult heart, may account for the observed degree of regeneration, but this has not been tested. We surveyed 120 inbred mouse strains and found that the frequency of adult mononuclear cardiomyocytes was surprisingly variable (>7-fold). Cardiomyocyte proliferation and heart functional recovery after coronary artery ligation both correlated with pre-injury MNDCM content. Using genome-wide association, we identified Tnni3k as one gene that influences variation in this composition and demonstrated that Tnni3k knockout resulted in elevated MNDCM content and increased cardiomyocyte proliferation after injury. Reciprocally, overexpression of Tnni3k in zebrafish promoted cardiomyocyte polyploidization and compromised heart regeneration. Our results corroborate the relevance of MNDCMs in heart regeneration. Moreover, they imply that intrinsic heart regeneration is not limited nor uniform in all individuals, but rather is a variable trait influenced by multiple genes.
Sigma-Aldrich
Nuclei Isolation Kit: Nuclei PURE Prep, sufficient for 15 nuclei preparations (~1-10×107 cells or 1g of tissue per preparation)
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit