Merck
  • Determination of the operational molarity of solutions of bovine alpha-chymotrypsin, trypsin, thrombin and factor Xa by spectrofluorimetric titration.
Determination of the operational molarity of solutions of bovine alpha-chymotrypsin, trypsin, thrombin and factor Xa by spectrofluorimetric titration.
The Biochemical journal (1973-01-01)
G W Jameson, D V Roberts, R W Adams, W S Kyle, D T Elmore
ABSTRACT
Several esters of 4-methylumbelliferone and 2-naphthol were synthesized and examined as possible spectrofluorimetric titrants for bovine alpha-chymotrypsin, trypsin, thrombin, Factor Xa and for subtilisin Novo. 4-Methylumbelliferyl p-guanidinobenzoate hydrochloride (MUGB) is a satisfactory titrant for alpha- and beta-trypsin, thrombin and Factor Xa and 4-methylumbelliferyl p-(NNN-trimethylammonium)cinnamate (MUTMAC) is a good titrant for alpha-chymotrypsin. The amount of enzyme used for spectrofluorimetric titration is 0.02-3.00nmol and the amount of 4-methylumbelliferone liberated is independent of the concentration of titrant and stoicheiometrically equal to the amount of enzyme used. Results obtained with MUGB and MUTMAC have been checked by spectrophotometric titration with p'-nitrophenyl p-guanidinobenzoate hydrochloride and p-nitrophenyl N(2)-acetyl-N(1)-benzylcarbazate respectively. p-Nitrophenyl N(2)-acetyl-N(1)-(9-anthrylmethyl)carbazate has been synthesized; it did not react with alpha-chymotrypsin. A satisfactory spectrofluorimetric titrant for subtilisin Novo was not discovered.