Merck
Regulation of Autophagic Flux by the 20S Proteasome.
Cell chemical biology (2019-07-23)
Evert Njomen, Jetze J Tepe
ABSTRACT
The proteolytic arm of the protein homeostasis network is maintained by both the ubiquitin-proteasome system (UPS) and autophagy. A well-balanced crosstalk between the two catabolic pathways ensures energy-efficient maintenance of cellular function. Our current understanding of the crosstalk between the UPS and autophagy is centered around substrate ubiquitination. Herein we report an additional method of crosstalk involving ubiquitin-independent 20S proteasome regulation of autophagosome-lysosome fusion. We found that enhancement of 20S proteasome activity increased the degradation of the disordered soluble N-ethylmaleimide-sensitive factor activating protein receptor proteins, synaptosomal-associated protein 29 (SNAP29), and syntaxin 17 (STX17), but not vesicle-associated membrane protein 8. This resulted in a reduction of autophagosome-lysosome fusion, which was ameliorated upon overexpression of both SNAP29 and STX17. In all, we herein present a mechanism of crosstalk between the proteasome and autophagy pathway that is regulated by ubiquitin-independent 20S proteasome-mediated degradation of SNAP29 and STX17.
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
TCH-165, ≥98% (HPLC)
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Tablets, For General Use
Sigma-Aldrich
MISSION® esiRNA, targeting human PSMD2
Sigma-Aldrich
N-Succinyl-Leu-Leu-Val-Tyr-7-Amido-4-Methylcoumarin, ≥90% (HPLC)
Sigma-Aldrich
Anti -Dinitrophenyl antibody, Mouse monoclonal, IgE isotype, ~1 mg/mL, clone SPE-7, affinity purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)