Merck
  • Ablation of BMP signaling hampers the blastema formation in Poecilia latipinna by dysregulating the extracellular matrix remodeling and cell cycle turnover.
Ablation of BMP signaling hampers the blastema formation in Poecilia latipinna by dysregulating the extracellular matrix remodeling and cell cycle turnover.
Zoology (Jena, Germany) (2019-04-14)
Sonam Patel, Isha Ranadive, Shailja Rajaram, Isha Desai, Suresh Balakrishnan
ABSTRACT
Bone morphogenetic proteins play a pivotal role in the epimorphic regeneration in vertebrates. Blastema formation is central to the epimorphic regeneration and crucially determines its fate. Despite an elaborate understanding of importance of Bone morphogenetic protein signaling in regeneration, its specific role during the blastema formation remains to be addressed. Regulatory role of BMP signaling during blastema formation was investigated using LDN193189, a potent inhibitor of BMP receptors. The study involved morphological observation, in vivo proliferation assay by incorporation of BrdU, comet assay, qRT-PCR and western blot. Blastemal outgrowth was seen reduced due to LDN193189 treatment, typified by dimensional differences, reduced number of proliferating cells and decreased levels of PCNA. Additionally, proapoptotic markers were found to be upregulated signifying a skewed cellular turnover. Further, the cell migration was seen obstructed and ECM remodeling was disturbed as well. These findings were marked by differential transcript as well as protein expressions of the key signaling and regulatory components, their altered enzymatic activities and other microscopic as well as molecular characterizations. Our results signify, for the first time, that BMP signaling manifests its effect on blastema formation by controlling the pivotal cellular processes possibly via PI3K/AKT. Our results indicate the pleiotropic role of BMPs specifically during blastema formation in regulating cell migration, cell proliferation and apoptosis, and lead to the generation of a molecular regulatory map of determinative molecules.
Product Description
Roche
Streptavidin-AP conjugate, solution, pkg of 1 mL (1,000U)