- Review article: treatment options for functional dyspepsia.
Review article: treatment options for functional dyspepsia.
Functional dyspepsia, consisting of epigastric pain syndrome and postprandial distress syndrome, is a prevalent functional gastrointestinal disorder. To date, only limited treatment options are available and conflicting results in terms of efficacy have been reported. Consequently, nonpharmacological treatment options are increasingly being explored for functional dyspepsia. To provide an overview of current pharmacological and nonpharmacological treatment options for functional dyspepsia. A literature search was conducted on Pubmed and other sources to identify relevant studies. Acid suppressive therapy reduced symptoms in 30%-70% of the patients, with higher benefit in epigastric pain syndrome and superior effectiveness for proton pump inhibitors compared to H2 -antagonists. Prokinetic agents, primarily used to treat postprandial distress syndrome, showed variable efficiency: 59%-81% responder rate for dopamine receptor antagonists, 32%-91% for serotonin-4-receptor agonists and 31%-80% for muscarinic receptor antagonists. H Pylori eradication, recommended in infected patients, was effective in 24%-82%. Refractory symptoms are addressed with neuromodulators. However, their efficacy in functional dyspepsia remains incompletely elucidated, available data showing symptom reduction in 27%-71% of the patients. Regarding herbal agents, peppermint oil reduced symptoms in 66%-91%, rikkunshito in 29%-34% and iberogast in 20%-95%. Lastly, acupuncture, cognitive behavioural therapy and hypnotherapy may help to provide symptom control, but research on their efficacy remains sparse. None of the available therapies is effective in the majority of patients without being associated with major side effects. Developing new treatment options is challenging due to the heterogeneity of functional dyspepsia, the lack of readily identified target mechanisms and the poor association between pathophysiological disturbances and symptoms.