- The Potential Chondrogenic Effect of Eucomis autumnalis Aqueous Extracts on Porcine Adipose-Derived Mesenchymal Stem Cells.
The Potential Chondrogenic Effect of Eucomis autumnalis Aqueous Extracts on Porcine Adipose-Derived Mesenchymal Stem Cells.
Tissue engineering. Part A (2018-12-05)
Makwese Maepa, Cornelius Cano Ssemakalu, Keolebogile Shirley Motaung
PMID30511903
ABSTRACT
Eucomis autumnalis is one plant that is used by various traditional healers to alleviate the signs and symptoms associated with osteoarthritis. Although the exact mechanisms remain unknown, we hypothesized that this plant can induce chondrogenesis. In this work, we explored the potential for an aqueous crude extract from E. autumnalis to induce chondrogenesis in porcine adipose-derived mesenchymal stem cells (MSCs). The results reported in our article indicate that the aqueous crude extract from E. autumnalis was able to indeed induce chondrogenesis. Our research is relevant to communities that rely on plant-based remedies for their well-being.