Merck
  • Profiling of polar metabolites in fruits of Opuntia stricta var. dillenii by ion-pair high-performance countercurrent chromatography and off-line electrospray mass-spectrometry injection.
Profiling of polar metabolites in fruits of Opuntia stricta var. dillenii by ion-pair high-performance countercurrent chromatography and off-line electrospray mass-spectrometry injection.
Journal of chromatography. A (2019-06-20)
Thu Minh Thi Tran, Binh Nguyen Thanh, Tamer E Moussa-Ayoub, Sascha Rohn, Gerold Jerz
ABSTRACT
Electrospray mass spectrometry-profiling guided the metabolome investigation of a C18 reversed phase adsorbate of Opuntia stricta var. dillenii fruits following analytical, and semi-preparative high-performance countercurrent chromatography (HPCCC) fractionation, and visualization of molecular weight elution profiles based on selected single ion-traces. Experimental partition-ratio values KD, and peak widths for detected metabolites were determined. Structural characterization of metabolites, and co-elution effects were monitored in the scan range m/z 150-2200. The polar ion-pair activated solvent system tert.-butylmethylether - n-butanol - acetonitrile - water (0.7% trifluoroacetic acid) [2:2:1:5]) was used for partition-ratio KD improvement of ionic betalains. HPCCC operations were in the head-to-tail mode using the elution-extrusion approach. Selected ESI-MS ion traces visualized the elution of fourty-three metabolites, whereas twenty-one were identified as known betalain pigments, and their classic degradation products, and chlorinated betacyanin artefacts. Potentially, novel fruit metabolites of Opuntia were recognized by unknown molecular weights, and MS/MS fragmentations. Off-line ESI-MS fraction monitoring determined peak elution windows, and resulted in KD-based chromatographic scales. Detectable metabolites were compared by separation- α, and resolution-factors RS revealing a better performance of the analytical HPCCC experiment. Experimental metabolite KD-values from analytical and semi-preparative HPCCC runs were widely consistent, and confirmed the reproducibility of the technique based on the used sample material.