Merck
  • Hypoglycaemic effects of glimepiride in sulfonylurea receptor 1 deficient rat.
Hypoglycaemic effects of glimepiride in sulfonylurea receptor 1 deficient rat.
British journal of pharmacology (2018-11-25)
Xiaojun Zhou, Rui Zhang, Zhiwei Zou, Xue Shen, Tianyue Xie, Chunmei Xu, Jianjun Dong, Lin Liao
ABSTRACT
Sulfonylureas (SUs) have been suggested to have an insulin-independent blood glucose-decreasing activity due to an extrapancreatic effect. However, a lack of adequate in vivo evidence makes this statement controversial. Here, we aimed to evaluate whether glimepiride has extrapancreatic blood glucose-lowering activity in vivo. Sulfonylurea receptor 1 deficient (SUR1-/- ) rats were created by means of transcription activator-like effector nucleases (TALEN)-mediated gene targeting technology. Type 2 diabetic models were established by feeding a high-fat diet and administering a low-dose of streptozotocin. These rats were then randomly divided into four groups: glimepiride, gliclazide, metformin and saline. All rats were treated for 2 weeks. Glimepiride decreased blood glucose levels and increased insulin sensitivity without elevating insulin levels. Gliclazide showed similar effects as glimepiride. Both agents were weaker than metformin. Further mechanistic investigations revealed that glimepiride increased hepatic glycogen synthesis and decreased gluconeogenesis, which were accompanied by the activation of Akt in the liver. Moreover, glimepiride increased both total and membrane glucose transporter 4 (GLUT4) levels in muscle and fat, which might be attributed to insulin receptor-independent IRS1/Akt activation. Glimepiride possesses an extrapancreatic blood glucose-lowering effect in vivo, which might be attributed to its direct effect on insulin-sensitive tissues. Therefore, the combination of glimepiride with multiple insulin injections should not be excluded per se.
Sigma-Aldrich
Anti-phospho-Insulin Receptor Substrate-1 (pTyr612) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution