Merck
  • Translating peripheral bladder afferent mechanosensitivity to neuronal activation within the lumbosacral spinal cord of mice.
Translating peripheral bladder afferent mechanosensitivity to neuronal activation within the lumbosacral spinal cord of mice.
Pain (2018-12-12)
Luke Grundy, Andrea M Harrington, Ashlee Caldwell, Joel Castro, Vasiliki Staikopoulos, Vladimir P Zagorodnyuk, Simon J H Brookes, Nick J Spencer, Stuart M Brierley
ABSTRACT
Primary afferent neurons transduce distension of the bladder wall into action potentials that are relayed into the spinal cord and brain, where autonomic reflexes necessary for maintaining continence are coordinated with pathways involved in sensation. However, the relationship between spinal circuits involved with physiological and nociceptive signalling from the bladder has only been partially characterised. We used ex vivo bladder afferent recordings to characterise mechanosensitive afferent responses to graded distension (0-60 mm Hg) and retrograde tracing from the bladder wall to identify central axon projections within the dorsal horn of the lumbosacral (LS) spinal cord. Labelling of dorsal horn neurons with phosphorylated-MAP-kinase (pERK), combined with labelling for neurochemical markers (calbindin, calretinin, gamma aminobutyric acid, and parvalbumin) after in vivo bladder distension (20-60 mm Hg), was used to identify spinal cord circuits processing bladder afferent input. Ex vivo bladder distension evoked an increase in primary afferent output, and the recruitment of both low- and high-threshold mechanosensitive afferents. Retrograde tracing revealed bladder afferent projections that localised with pERK-immunoreactive dorsal horn neurons within the superficial laminae (superficial dorsal horn), dorsal gray commissure, and lateral collateral tracts of the LS spinal cord. Populations of pERK-immunoreactive neurons colabelled with calbindin, calretinin, or gamma aminobutyric acid, but not parvalbumin. Noxious bladder distension increased the percentage of pERK-immunoreactive neurons colabelled with calretinin. We identified LS spinal circuits supporting autonomic and nociceptive reflexes responsible for maintaining continence and bladder sensations. Our findings show for the first time that low- and high-threshold bladder afferents relay into similar dorsal horn circuits, with nociceptive signalling recruiting a larger number of neurons.