Merck
  • New Noonan syndrome model mice with RIT1 mutation exhibit cardiac hypertrophy and susceptibility to β-adrenergic stimulation-induced cardiac fibrosis.
New Noonan syndrome model mice with RIT1 mutation exhibit cardiac hypertrophy and susceptibility to β-adrenergic stimulation-induced cardiac fibrosis.
EBioMedicine (2019-03-23)
Shingo Takahara, Shin-Ichi Inoue, Sachiko Miyagawa-Tomita, Katsuhisa Matsuura, Yasumi Nakashima, Tetsuya Niihori, Yoichi Matsubara, Yoshikatsu Saiki, Yoko Aoki
ABSTRACT
Noonan syndrome (NS) is a genetic disorder characterized by short stature, a distinctive facial appearance, and heart defects. We recently discovered a novel NS gene, RIT1, which is a member of the RAS subfamily of small GTPases. NS patients with RIT1 mutations have a high incidence of hypertrophic cardiomyopathy and edematous phenotype, but the specific role of RIT1 remains unclear. To investigate how germline RIT1 mutations cause NS, we generated knock-in mice that carried a NS-associated Rit1 A57G mutation (Rit1A57G/+). We investigated the phenotypes of Rit1A57G/+ mice in fetal and adult stages as well as the effects of isoproterenol on cardiac function in Rit1A57G/+ mice. Rit1A57G/+ embryos exhibited decreased viability, edema, subcutaneous hemorrhage and AKT activation. Surviving Rit1A57G/+ mice had a short stature, craniofacial abnormalities and splenomegaly. Cardiac hypertrophy and cardiac fibrosis with increased expression of S100A4, vimentin and periostin were observed in Rit1A57G/+ mice compared to Rit1+/+ mice. Upon isoproterenol stimulation, cardiac fibrosis was drastically increased in Rit1A57G/+ mice. Phosphorylated (at Thr308) AKT levels were also elevated in isoproterenol-treated Rit1A57G/+ hearts. The A57G mutation in Rit1 causes cardiac hypertrophy, fibrosis and other NS-associated features. Biochemical analysis indicates that the AKT signaling pathway might be related to downstream signaling in the RIT1 A57G mutant at a developmental stage and under β-adrenergic stimulation in the heart. FUND: The Grants-in-Aid were provided by the Practical Research Project for Rare/Intractable Diseases from the Japan Agency for Medical Research and Development, the Japan Society for the Promotion of Science KAKENHI Grant.
Sigma-Aldrich
Anti-RIT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution