Merck
  • On the mechanism of thrombin-induced angiogenesis: involvement of alphavbeta3-integrin.
On the mechanism of thrombin-induced angiogenesis: involvement of alphavbeta3-integrin.
American journal of physiology. Cell physiology (2002-10-10)
Nikos E Tsopanoglou, Paraskevi Andriopoulou, Michael E Maragoudakis
ABSTRACT
Thrombin has been reported to be a potent angiogenic factor both in vitro and in vivo, and many of the cellular effects of thrombin may contribute to activation of angiogenesis. In this report we show that thrombin-treatment of human endothelial cells increases mRNA and protein levels of alphavbeta3-integrin. This thrombin-mediated effect is specific, dose dependent, and requires the catalytic site of thrombin. In addition, thrombin interacts with alphavbeta3 as demonstrated by direct binding of alphavbeta3 protein to immobilized thrombin. This interaction of thrombin with alphavbeta3-integrin, which is an angiogenic marker in vascular tissue, is of functional significance. Immobilized thrombin promotes endothelial cells attachment, migration, and survival. Antibody to alphavbeta3 or a specific peptide antagonist to alphavbeta3 can abolish all these alphavbeta3-mediated effects. Furthermore, in the chick chorioallantoic membrane system, the antagonist peptide to alphavbeta3 diminishes both basal and the thrombin-induced angiogenesis. These results support the pivotal role of thrombin in activation of endothelial cells and angiogenesis and may be related to the clinical observation of neovascularization within thrombi.
Sigma-Aldrich
Anti-Integrin β3 Antibody, clone PM6/13, clone PM6/13, Chemicon®, from mouse
Sigma-Aldrich
Human Integrin αVβ3 Protein, Triton X-100 Formulation