Merck
  • Digital microfluidic immobilized cytochrome P450 reactors with integrated inkjet-printed microheaters for droplet-based drug metabolism research.
Digital microfluidic immobilized cytochrome P450 reactors with integrated inkjet-printed microheaters for droplet-based drug metabolism research.
Analytical and bioanalytical chemistry (2018-08-04)
Gowtham Sathyanarayanan, Markus Haapala, Iiro Kiiski, Tiina Sikanen
ABSTRACT
We report the development and characterization of digital microfluidic (DMF) immobilized enzyme reactors (IMERs) for studying cytochrome P450 (CYP)-mediated drug metabolism on droplet scale. The on-chip IMERs consist of porous polymer (thiol-ene) monolith plugs prepared in situ by photopolymerization and functionalized with recombinant CYP1A1 isoforms (an important detoxification route for many drugs and other xenobiotics). The DMF devices also incorporate inexpensive, inkjet-printed microheaters for on-demand regio-specific heating of the IMERs to physiological temperature, which is crucial for maintaining the activity of the temperature-sensitive CYP reaction. For on-chip monitoring of the CYP activity, the DMF devices were combined with a commercial well-plate reader, and a custom fluorescence quantification method was developed for detection of the chosen CYP1A1 model activity (ethoxyresorufin-O-deethylation). The reproducibility of the developed assay was examined with the help of ten parallel CYP-IMERs. All CYP-IMERs provided statistically significant difference (in fluorescence response) compared to any of the negative controls (including room-temperature reactions). The average (n = 10) turnover rate was 20.3 ± 9.0 fmol resorufin per minute. Via parallelization, the concept of the droplet-based CYP-IMER developed in this study provides a viable approach to rapid and low-cost prediction of the metabolic clearance of new chemical entities in vitro.
Sigma-Aldrich
Pentaerythritol tetrakis(3-mercaptopropionate), >95%