- Coregulator-related diseases.
Coregulator-related diseases.
Internal medicine (Tokyo, Japan) (2004-06-23)
Toshihiko Yanase, Masahiro Adachi, Kiminobu Goto, Ryoichi Takayanagi, Hajime Nawata
PMID15206547
ABSTRACT
Coregulators are a group of proteins, which modulate the nuclear receptor transactivation function. In this study, a new "coregulator disease" concept was proposed from observations of a case of androgen insensitivity syndrome (AIS) and cases involving Rubinstein-Taybi syndrome and X-linked dementia and hypothyroidism syndrome. In addition, coregulators are thought to be closely associated with the pathogenesis of several diseases such as hormone-dependent cancers and leukemia. Based on these observations, the clinical disorders associated with some coregulator abnormalities were reviewed.
Sigma-Aldrich
SRC1 (627-786), biotin,His tagged human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
SRC1, His tagged, TEV site, human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
SRC1, FLAG tagged, TEV site human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)