Merck
Phosphorylated CREB binds specifically to the nuclear protein CBP.
Nature (1993-10-28)
J C Chrivia, R P Kwok, N Lamb, M Hagiwara, M R Montminy, R H Goodman
ABSTRACT
Cyclic AMP-regulated gene expression frequently involves a DNA element known as the cAMP-regulated enhancer (CRE). Many transcription factors bind to this element, including the protein CREB, which is activated as a result of phosphorylation by protein kinase A. This modification stimulates interaction with one or more of the general transcription factors or, alternatively, allows recruitment of a co-activator. Here we report that CREB phosphorylated by protein kinase A binds specifically to a nuclear protein of M(r) 265K which we term CBP (for CREB-binding protein). Fusion of a heterologous DNA-binding domain to the amino terminus of CBP enables the chimaeric protein to function as a protein kinase A-regulated transcriptional activator. We propose that CBP may participate in cAMP-regulated gene expression by interacting with the activated phosphorylated form of CREB.
Sigma-Aldrich
p300 (C1135-2414) human, recombinant, expressed in insect cells, ≥70% (SDS-PAGE)