Merck
  • Detection of Total Ergot Alkaloids in Cereal Flour and in Bread by a Generic Enzyme Immunoassay Method.
Detection of Total Ergot Alkaloids in Cereal Flour and in Bread by a Generic Enzyme Immunoassay Method.
Journal of AOAC International (2017-10-02)
Madeleine Gross, Valeriu Curtui, Ewald Usleber
ABSTRACT
Four sets of polyclonal antibodies against ergot alkaloids ergometrine, ergotamine, α-ergocryptine, and ergocornine were produced and characterized in a competitive direct or indirect enzyme immunoassay (EIA). Standard curve LODs were 0.03 ng/mL (ergometrine EIA) to 2.0 ng/mL (ergocornine EIA). Three EIAs were highly specific, whereas the ergometrine EIA had a broad specificity pattern and reacted, albeit weakly, with all seven major ergot alkaloids and their epimeric forms. Using the ergometrine EIA, a generic test system was established in which total ergot alkaloids are quantified by a standard curve for a toxin mixture composed of three alkaloids that matched the ergot alkaloid composition in naturally contaminated rye and wheat products. Sample extraction with acetonitrile-phosphate-buffered saline at pH 6.0 without further cleanup was sufficient for EIA analysis. The LODs for total ergot alkaloids were 20 ng/g in rye and wheat flour and 14 ng/g in bread. Recoveries were 85-110% (RSDs of 0.1-11.7%) at a concentration range of 50-1000 ng/g. The total ergot alkaloid EIA was validated by comparison with HPLC-fluorescence detection. Although some under- and overestimation by the total ergot alkaloid EIA was observed, it was suitable for the reliable identification of positive samples at 10-20 ng/g and for the determination of total ergot alkaloids in a concentration range between 100 and 1000 ng/g.