Merck
  • Synthesis, Characterization, and Biological Evaluation of Unimetallic and Heterobimetallic Complexes of Bivalent Copper.
Synthesis, Characterization, and Biological Evaluation of Unimetallic and Heterobimetallic Complexes of Bivalent Copper.
Bioinorganic chemistry and applications (2018-06-01)
Anshul Singh, Ashu Chaudhary
ABSTRACT
We present an inclusive characterization of the unimetallic and heterobimetallic complexes of copper synthesized using CuCl2 and diamine (4-fluoro 1,2-phenylenediamine) resulting in monometallic complex which further undergoes treatment with organometallic dichlorides of group 4 and 14 in 1 : 2 molar ratio resulting in heterobimetallic complexes. These complexes thoroughly characterized using various physical, analytical, and spectroscopic techniques indicate square planar and distorted octahedral geometry for the synthesized unimetallic and heterobimetallic complexes, respectively. These complexes were evaluated for their antimicrobial efficacy against various bacterial and fungal strains while hepatoprotective activity was also examined in male albino rats.
Sigma-Aldrich
4-Fluoro-1,2-phenylenediamine, 97%