![Zalcitabine pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/161/807/0a629238-8f3d-45d6-a674-9b4730bf8f20/640/0a629238-8f3d-45d6-a674-9b4730bf8f20.png)
Zalcitabine
![Zalcitabine pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/161/807/0a629238-8f3d-45d6-a674-9b4730bf8f20/640/0a629238-8f3d-45d6-a674-9b4730bf8f20.png)
Synonym(s):
2′,3′-Dideoxycytidine, ddC
Empirical Formula (Hill Notation):
C9H13N3O3
CAS Number:
Molecular Weight:
211.22
Products
Brand
Formula Weight
Melting Point (°C)
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing